The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165927.1 | NP_001159399.1 | 8 - 9 | 927 | 58 |
NM_001321268.1 | NP_001308197.1 | 7 - 8 | 840 | 58 | |
NM_001321269.1 | NP_001308198.1 | 8 - 9 | 938 | 58 | |
NM_017777.3 | NP_060247.2 | 8 - 9 | 938 | 58 | |
XM_005257485.3 | XP_005257542.1 | 8 - 9 | 919 | 58 | |
XM_006721965.2 | XP_006722028.1 | 7 - 8 | 772 | 58 | |
XM_011524957.2 | XP_011523259.1 | 8 - 9 | 918 | 58 | |
XM_011524958.2 | XP_011523260.1 | 8 - 9 | 918 | 58 | |
XM_011524959.2 | XP_011523261.1 | 8 - 9 | 917 | 58 | |
XM_011524960.2 | XP_011523262.1 | 8 - 9 | 917 | 58 | |
XM_017024803.1 | XP_016880292.1 | 8 - 9 | 886 | 58 | |
XM_017024804.1 | XP_016880293.1 | 8 - 9 | 886 | 58 | |
XM_017024805.1 | XP_016880294.1 | 8 - 9 | 952 | 58 | |
GenBank mRNA | AK000352.1 | - | 5 - 6 | 608 | 58 |
AK301020.1 | - | 7 - 8 | 771 | 58 | |
AK310815.1 | - | 8 - 9 | 927 | 58 | |
BC010061.2 | - | 8 - 9 | 873 | 58 | |
CR457229.1 | - | 4 - 5 | 434 | 58 | |
DQ185029.1 | - | 8 - 9 | 938 | 58 | |
KU178661.1 | - | 4 - 5 | 434 | 58 | |
KU178662.1 | - | 4 - 5 | 434 | 58 | |
KU178663.1 | - | 4 - 5 | 434 | 58 |