Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CDK10
- Entrez Gene ID
-
8558
- Gene Name
- cyclin dependent kinase 10
- Gene Aliases
-
PISSLRE
- Chromosome Location
-
Chr.16: 89686612 - 89696364 on Build GRCh38
View Full Product Details