The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320772.1 | NP_001307701.1 | 9 - 10 | 1093 | 96 |
NM_017983.6 | NP_060453.3 | 10 - 11 | 1176 | 96 | |
NR_135470.1 | 10 - 11 | 1312 | 96 | ||
NR_135471.1 | 9 - 10 | 1105 | 96 | ||
XM_017024808.1 | XP_016880297.1 | 8 - 9 | 3130 | 96 | |
GenBank mRNA | AK301830.1 | - | 9 - 10 | 1039 | 96 |
AY691424.1 | - | 10 - 11 | 1076 | 96 | |
BC039867.2 | - | 10 - 11 | 1120 | 96 |