The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301627.1 | NP_001288556.1 | 9 - 10 | 1494 | 93 |
NM_052917.3 | NP_443149.2 | 11 - 12 | 1972 | 93 | |
XM_011510537.2 | XP_011508839.1 | 11 - 12 | 1967 | 93 | |
XM_011510538.2 | XP_011508840.1 | 9 - 10 | 1379 | 93 | |
XM_017003258.1 | XP_016858747.1 | 11 - 12 | 1821 | 93 | |
XM_017003259.1 | XP_016858748.1 | 10 - 11 | 1546 | 93 | |
XM_017003260.1 | XP_016858749.1 | 11 - 12 | 1967 | 93 | |
XM_017003261.1 | XP_016858750.1 | 11 - 12 | 1831 | 93 | |
XM_017003262.1 | XP_016858751.1 | 9 - 10 | 1237 | 93 | |
XM_017003263.1 | XP_016858752.1 | 9 - 10 | 1379 | 93 | |
GenBank mRNA | AB067505.1 | - | 9 - 10 | 1272 | 93 |
AB078142.1 | - | 9 - 10 | 1390 | 93 | |
AJ505991.2 | - | 9 - 10 | 1390 | 93 | |
AK131195.1 | - | 11 - 12 | 1917 | 93 | |
BC101031.1 | - | 9 - 10 | 1390 | 93 | |
BC101032.2 | - | 9 - 10 | 1390 | 93 | |
BC101033.1 | - | 9 - 10 | 1390 | 93 | |
BC101034.2 | - | 9 - 10 | 1390 | 93 | |
KJ534843.1 | - | 9 - 10 | 1390 | 93 | |
KJ535058.1 | - | 8 - 9 | 1221 | 93 |