The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278352.1 | NP_001265281.1 | 15 - 16 | 1949 | 64 |
NM_002197.2 | NP_002188.1 | 14 - 15 | 1877 | 64 | |
XM_005251476.1 | XP_005251533.1 | 15 - 16 | 1887 | 64 | |
XM_011517888.1 | XP_011516190.1 | 14 - 15 | 1799 | 64 | |
GenBank mRNA | AB209480.1 | - | 14 - 15 | 1832 | 64 |
AF261088.1 | - | 15 - 16 | 1928 | 64 | |
AK308752.1 | - | 13 - 14 | 1843 | 64 | |
BC018103.1 | - | 13 - 14 | 1759 | 64 | |
EU794627.1 | - | 13 - 14 | 1759 | 64 | |
M58510.1 | - | 12 - 13 | 1505 | 64 | |
Z11559.1 | - | 14 - 15 | 1829 | 64 |