The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278352.1 | NP_001265281.1 | 19 - 20 | 2474 | 65 |
NM_002197.2 | NP_002188.1 | 18 - 19 | 2402 | 65 | |
XM_005251476.1 | XP_005251533.1 | 19 - 20 | 2412 | 65 | |
XM_011517888.1 | XP_011516190.1 | 18 - 19 | 2324 | 65 | |
GenBank mRNA | AB209480.1 | - | 18 - 19 | 2603 | 65 |
AF261088.1 | - | 19 - 20 | 2453 | 65 | |
BC018103.1 | - | 17 - 18 | 2284 | 65 | |
EU794627.1 | - | 17 - 18 | 2284 | 65 | |
M58510.1 | - | 16 - 17 | 2030 | 65 | |
Z11559.1 | - | 18 - 19 | 2354 | 65 |