Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDDC1
Entrez Gene ID
347862
Gene Name
Parkinson disease 7 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.11: 767095 - 777502 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318818.1 NP_001305747.1 4 - 5 409 55
NM_001318820.1 NP_001305749.1 4 - 5 378 55
NM_001318821.1 NP_001305750.1 5 - 6 486 55
NM_001318822.1 NP_001305751.1 3 - 4 301 55
NM_001318823.1 NP_001305752.1 4 - 5 409 55
NM_001318824.1 NP_001305753.1 4 - 5 378 55
NM_182612.3 NP_872418.1 5 - 6 486 55
NR_134867.1 4 - 5 409 55
NR_134868.1 5 - 6 580 55
XM_011520064.1 XP_011518366.1 5 - 6 468 55
XM_017017660.1 XP_016873149.1 5 - 6 471 55
XM_017017661.1 XP_016873150.1 5 - 6 468 55
XM_017017663.1 XP_016873152.1 4 - 5 371 55
XM_017017666.1 XP_016873155.1 4 - 5 392 55
XM_017017668.1 XP_016873157.1 4 - 5 392 55
GenBank mRNA AK092816.1 - 4 - 5 363 55
AK293559.1 - 4 - 5 394 55
AK309153.1 - 4 - 5 392 55
BC110868.1 - 5 - 6 455 55
BC112145.1 - 5 - 6 467 55
BC143415.1 - 3 - 4 282 55
BC143416.1 - 4 - 5 359 55
BC143418.1 - 5 - 6 467 55
BC143419.1 - 4 - 5 390 55
BC143420.1 - 4 - 5 359 55
BC143421.1 - 5 - 6 561 55
BC143422.1 - 4 - 5 390 55
BC143423.1 - 4 - 5 390 55
DA025348.1 - 5 - 6 471 55
DC364167.1 - 5 - 6 486 55

View Full Product Details