Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LINC00470
Entrez Gene ID
56651
Gene Name
long intergenic non-protein coding RNA 470
Gene Aliases
C18orf2
Chromosome Location
Chr.18: 1268311 - 1359629 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_023925.1 1 - 2 244 207
NR_023926.1 1 - 2 244 161
NR_110327.1 1 - 2 244 151
GenBank mRNA AF295725.1 - 1 - 2 244 207
AF295726.1 - 1 - 2 205 161


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AA021221.1 - 1 - 2 173 Chr. 18 - 1269796 - 1359554 [-] on Build GRCh38 151
GenBankmRNA AA013326.1 - 172 Chr. - - [-] on Build 151
GenBankmRNA AA019670.1 - 1 - 2 172 Chr. 18 - 1275988 - 1359557 [-] on Build GRCh38 152
GenBankmRNA AA057462.1 - 1 - 2 172 Chr. 18 - 1269617 - 1359557 [-] on Build GRCh38 151
GenBankmRNA AA056149.1 - 1 - 2 172 Chr. 18 - 1269660 - 1359557 [-] on Build GRCh38 151
GenBankmRNA W92284.1 - 173 Chr. - - [-] on Build 151
GenBankmRNA H85330.1 - 1 - 2 186 Chr. 18 - 1272452 - 1359560 [-] on Build GRCh38 151


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream uncharacterized LOC388456 LOC388456 -430.7kb Sense Chr. - 926083 - 928129 on Build GRCh38
Downstream uncharacterized LOC101926997 LOC101926997 1.2mb Sense Chr. - 2517250 - 2519260 on Build GRCh38

View Full Product Details