Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLSCR4
Entrez Gene ID
57088
Gene Name
phospholipid scramblase 4
Gene Aliases
TRA1
Chromosome Location
Chr.3: 146192335 - 146251179 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128304.1 NP_001121776.1 8 - 9 997 84
NM_001128305.1 NP_001121777.1 6 - 7 959 84
NM_020353.2 NP_065086.2 6 - 7 871 84
XM_005247654.2 XP_005247711.1 7 - 8 898 84
XM_005247655.2 XP_005247712.1 7 - 8 815 84
XM_011513031.2 XP_011511333.1 6 - 7 757 84
GenBank mRNA AF087887.1 - 3 - 4 504 84
AF199023.1 - 6 - 7 775 84
AK290214.1 - 6 - 7 899 84
AK292815.1 - 8 - 9 897 84
AK312809.1 - 6 - 7 813 84
AL833760.1 - 6 - 7 785 84
AY550971.1 - 6 - 7 725 84
BC028354.2 - 6 - 7 871 84

View Full Product Details