The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128304.1 | NP_001121776.1 | 10 - 11 | 1309 | 88 |
NM_001128305.1 | NP_001121777.1 | 8 - 9 | 1271 | 88 | |
NM_001128306.1 | NP_001121778.1 | 6 - 7 | 913 | 88 | |
NM_001177304.1 | NP_001170775.1 | 5 - 6 | 885 | 88 | |
NM_020353.2 | NP_065086.2 | 8 - 9 | 1183 | 88 | |
XM_005247654.2 | XP_005247711.1 | 9 - 10 | 1210 | 88 | |
XM_005247655.2 | XP_005247712.1 | 9 - 10 | 1127 | 88 | |
XM_011513031.2 | XP_011511333.1 | 8 - 9 | 1069 | 88 | |
XM_017006921.1 | XP_016862410.1 | 7 - 8 | 940 | 88 | |
GenBank mRNA | AF199023.1 | - | 8 - 9 | 1087 | 88 |
AK128442.1 | - | 5 - 6 | 785 | 88 | |
AK290214.1 | - | 8 - 9 | 1211 | 88 | |
AK312809.1 | - | 8 - 9 | 1125 | 88 | |
AL833760.1 | - | 8 - 9 | 1097 | 88 | |
AY550971.1 | - | 8 - 9 | 1037 | 88 | |
BC028354.2 | - | 8 - 9 | 1183 | 88 |