Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WHRN
Entrez Gene ID
25861
Gene Name
whirlin
Gene Aliases
CIP98, DFNB31, PDZD7B, USH2D, WI
Chromosome Location
Chr.9: 114402078 - 114505509 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083885.2 NP_001077354.2 6 - 7 908 93
NM_001173425.1 NP_001166896.1 6 - 7 2072 93
NM_015404.3 NP_056219.3 6 - 7 2072 93
XM_011518484.1 XP_011516786.1 6 - 7 2125 93
XM_011518485.1 XP_011516787.1 7 - 8 1973 93
XM_011518486.1 XP_011516788.1 6 - 7 2125 93
XM_011518487.1 XP_011516789.1 5 - 6 1999 93
XM_011518489.2 XP_011516791.1 6 - 7 2125 93
XM_011518491.2 XP_011516793.1 6 - 7 2125 93
XM_011518492.1 XP_011516794.1 6 - 7 2125 93
XM_011518494.2 XP_011516796.1 6 - 7 2125 93
XM_011518495.1 XP_011516797.1 3 - 4 377 93
GenBank mRNA AB040959.1 - 6 - 7 1586 93
AK022854.1 - 2 - 3 786 93
AL110228.1 - 6 - 7 866 93
AL557526.3 - 2 - 3 585 93
BC014524.1 - - 206 93
BC136416.1 - 6 - 7 1821 93
BC142614.1 - 6 - 7 2072 93
BC142684.1 - 6 - 7 2047 93

View Full Product Details