Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GATA3
Entrez Gene ID
2625
Gene Name
GATA binding protein 3
Gene Aliases
HDR, HDRS
Chromosome Location
Chr.10: 8045421 - 8075201 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005252442.2 XP_005252499.1 1 - 2 171 118
GenBank mRNA BM152193.1 - 1 - 2 177 118

View Full Product Details