The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270526.1 | NP_001257455.1 | 1 - 2 | 178 | 96 |
NM_001270527.1 | NP_001257456.1 | 1 - 2 | 178 | 96 | |
NM_004099.5 | NP_004090.4 | 1 - 2 | 178 | 96 | |
NM_198194.2 | NP_937837.1 | 1 - 2 | 178 | 96 | |
GenBank mRNA | AK302977.1 | - | 1 - 2 | 104 | 72 |
AK304518.1 | - | 1 - 2 | 122 | 96 | |
AK307999.1 | - | 1 - 2 | 122 | 96 | |
BC010703.1 | - | 1 - 2 | 100 | 96 | |
BG701899.1 | - | 1 - 2 | 131 | 96 | |
BI553227.1 | - | 1 - 2 | 107 | 96 | |
BM451470.1 | - | - | 142 | 96 | |
BP208523.1 | - | 1 - 2 | 105 | 96 | |
DC368197.1 | - | 1 - 2 | 178 | 96 | |
X60067.1 | - | 1 - 2 | 61 | 96 |