The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171137.1 | NP_001164608.1 | 5 - 6 | 802 | 113 |
NM_018387.4 | NP_060857.2 | 5 - 6 | 819 | 113 | |
NR_033234.1 | 4 - 5 | 674 | 113 | ||
XM_017014890.1 | XP_016870379.1 | 4 - 5 | 646 | 113 | |
XM_017014891.1 | XP_016870380.1 | 4 - 5 | 619 | 113 | |
XM_017014892.1 | XP_016870381.1 | 4 - 5 | 878 | 113 | |
XM_017014893.1 | XP_016870382.1 | 4 - 5 | 588 | 113 | |
XM_017014894.1 | XP_016870383.1 | 5 - 6 | 2190 | 113 | |
XM_017014895.1 | XP_016870384.1 | 5 - 6 | 2187 | 113 | |
XM_017014896.1 | XP_016870385.1 | 5 - 6 | 1007 | 113 | |
XM_017014897.1 | XP_016870386.1 | 4 - 5 | 646 | 113 | |
XM_017014898.1 | XP_016870387.1 | 4 - 5 | 646 | 113 | |
GenBank mRNA | AF333337.1 | - | 3 - 4 | 416 | 113 |
AK027890.1 | - | 4 - 5 | 736 | 113 | |
AL136866.1 | - | 5 - 6 | 802 | 113 | |
AL833947.1 | - | 4 - 5 | 674 | 113 | |
AL834150.1 | - | 4 - 5 | 674 | 113 | |
BC002693.2 | - | 2 - 3 | 218 | 113 | |
BC017732.1 | - | 5 - 6 | 819 | 113 | |
BC108731.1 | - | 4 - 5 | 711 | 113 | |
EU794683.1 | - | 5 - 6 | 819 | 113 | |
KU178696.1 | - | 2 - 3 | 389 | 113 | |
KU178697.1 | - | 2 - 3 | 389 | 113 |