The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016604.3 | NP_057688.2 | 10 - 11 | 3249 | 88 |
XM_005272018.4 | XP_005272075.1 | 9 - 10 | 2508 | 88 | |
XM_011543488.2 | XP_011541790.1 | 10 - 11 | 2964 | 88 | |
XM_011543489.2 | XP_011541791.1 | 10 - 11 | 3423 | 88 | |
XM_017009584.1 | XP_016865073.1 | 6 - 7 | 2369 | 88 | |
GenBank mRNA | AB029005.2 | - | 10 - 11 | 3129 | 88 |
AF251039.1 | - | 10 - 11 | 2761 | 88 | |
AF338242.1 | - | 10 - 11 | 3249 | 88 | |
AK304371.1 | - | 3 - 4 | 465 | 88 | |
BC000539.2 | - | 7 - 8 | 2502 | 88 | |
BC001202.1 | - | 2 - 3 | 160 | 88 | |
BC117693.1 | - | 6 - 7 | 2421 | 88 | |
BC146788.1 | - | 10 - 11 | 3130 | 88 |