The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016604.3 | NP_057688.2 | 3 - 4 | 678 | 71 |
XM_005272018.4 | XP_005272075.1 | 3 - 4 | 537 | 71 | |
XM_011543488.2 | XP_011541790.1 | 3 - 4 | 393 | 71 | |
XM_011543489.2 | XP_011541791.1 | 3 - 4 | 852 | 71 | |
GenBank mRNA | AB029005.2 | - | 3 - 4 | 558 | 71 |
AF251039.1 | - | 2 - 3 | 287 | 71 | |
AF338242.1 | - | 3 - 4 | 678 | 71 | |
BC073869.1 | - | 2 - 3 | 180 | 71 | |
BC146788.1 | - | 3 - 4 | 559 | 71 | |
BG390435.1 | - | 2 - 3 | 179 | 71 |