The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016604.3 | NP_057688.2 | 9 - 10 | 3036 | 100 |
XM_005272018.4 | XP_005272075.1 | 8 - 9 | 2295 | 100 | |
XM_011543488.2 | XP_011541790.1 | 9 - 10 | 2751 | 100 | |
XM_011543489.2 | XP_011541791.1 | 9 - 10 | 3210 | 100 | |
XM_017009584.1 | XP_016865073.1 | 5 - 6 | 2156 | 100 | |
GenBank mRNA | AB029005.2 | - | 9 - 10 | 2916 | 100 |
AF251039.1 | - | 9 - 10 | 2548 | 100 | |
AF338242.1 | - | 9 - 10 | 3036 | 100 | |
AK304371.1 | - | 2 - 3 | 252 | 100 | |
BC000539.2 | - | 6 - 7 | 2289 | 100 | |
BC117693.1 | - | 5 - 6 | 2208 | 100 | |
BC146788.1 | - | 9 - 10 | 2917 | 100 |