The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052958.2 | NP_443190.2 | 9 - 10 | 1615 | 103 |
XM_011517445.2 | XP_011515747.1 | 9 - 10 | 1578 | 103 | |
XM_011517446.2 | XP_011515748.1 | 9 - 10 | 1587 | 103 | |
XM_011517448.2 | XP_011515750.1 | 8 - 9 | 1458 | 103 | |
XM_017013019.1 | XP_016868508.1 | 8 - 9 | 1458 | 103 | |
GenBank mRNA | AB056652.1 | - | 8 - 9 | 2583 | 103 |
AK094191.1 | - | 3 - 4 | 416 | 103 | |
BC041961.1 | - | 9 - 10 | 1615 | 103 |