Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNK2
Entrez Gene ID
65268
Gene Name
WNK lysine deficient protein kinase 2
Gene Aliases
NY-CO-43, P/OKcl.13, PRKWNK2, SDCCAG43
Chromosome Location
Chr.9: 93184156 - 93327581 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282394.1 NP_001269323.1 10 - 11 2384 86
NM_006648.3 NP_006639.3 10 - 11 2384 86
XM_005252137.3 XP_005252194.1 11 - 12 2616 86
XM_005252140.2 XP_005252197.1 11 - 12 2616 86
XM_005252141.2 XP_005252198.1 11 - 12 2616 86
XM_005252144.2 XP_005252201.1 11 - 12 2616 86
XM_011518926.2 XP_011517228.1 11 - 12 2616 86
XM_011518927.2 XP_011517229.1 11 - 12 2616 86
XM_011518928.2 XP_011517230.1 11 - 12 2616 86
XM_011518929.2 XP_011517231.1 11 - 12 2616 86
XM_011518930.2 XP_011517232.1 11 - 12 2616 86
XM_011518931.2 XP_011517233.1 11 - 12 2616 86
XM_011518932.2 XP_011517234.1 11 - 12 2616 86
XM_011518935.2 XP_011517237.1 11 - 12 2616 86
XM_011518936.2 XP_011517238.1 11 - 12 2616 86
XM_011518937.2 XP_011517239.1 10 - 11 1924 86
XM_011518939.2 XP_011517241.1 10 - 11 1969 86
XM_011518940.2 XP_011517242.1 10 - 11 1971 86
XM_017015045.1 XP_016870534.1 11 - 12 2616 86
XM_017015046.1 XP_016870535.1 11 - 12 2616 86
XM_017015047.1 XP_016870536.1 11 - 12 2616 86
XM_017015048.1 XP_016870537.1 11 - 12 2616 86
XM_017015049.1 XP_016870538.1 11 - 12 2616 86
XM_017015050.1 XP_016870539.1 11 - 12 2616 86
XM_017015051.1 XP_016870540.1 11 - 12 2616 86
XM_017015052.1 XP_016870541.1 11 - 12 2616 86
XM_017015053.1 XP_016870542.1 11 - 12 2616 86
XM_017015054.1 XP_016870543.1 11 - 12 2616 86
XM_017015055.1 XP_016870544.1 11 - 12 2616 86
GenBank mRNA AB044546.1 - 9 - 10 2252 86
AB051547.2 - 10 - 11 2371 86
AJ242724.4 - 10 - 11 2384 86

View Full Product Details