Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNK2
Entrez Gene ID
65268
Gene Name
WNK lysine deficient protein kinase 2
Gene Aliases
NY-CO-43, P/OKcl.13, PRKWNK2, SDCCAG43
Chromosome Location
Chr.9: 93184156 - 93327581 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282394.1 NP_001269323.1 26 - 27 6320 70
NM_006648.3 NP_006639.3 25 - 26 6209 70
XM_005252137.3 XP_005252194.1 26 - 27 6441 70
XM_005252140.2 XP_005252197.1 26 - 27 6441 70
XM_005252141.2 XP_005252198.1 26 - 27 6441 70
XM_005252144.2 XP_005252201.1 26 - 27 6441 70
XM_011518926.2 XP_011517228.1 26 - 27 6441 70
XM_011518927.2 XP_011517229.1 26 - 27 6438 70
XM_011518928.2 XP_011517230.1 26 - 27 6387 70
XM_011518930.2 XP_011517232.1 26 - 27 6306 70
XM_011518932.2 XP_011517234.1 26 - 27 6441 70
XM_011518937.2 XP_011517239.1 25 - 26 5749 70
XM_011518939.2 XP_011517241.1 25 - 26 5794 70
XM_011518940.2 XP_011517242.1 25 - 26 5796 70
XM_017015045.1 XP_016870534.1 26 - 27 6438 70
XM_017015046.1 XP_016870535.1 26 - 27 6303 70
XM_017015047.1 XP_016870536.1 26 - 27 6387 70
XM_017015048.1 XP_016870537.1 26 - 27 6384 70
XM_017015051.1 XP_016870540.1 26 - 27 6306 70
GenBank mRNA AB051547.2 - 25 - 26 6196 70
AJ242724.4 - 26 - 27 6320 70
AK000694.1 - 11 - 12 2599 70
BC037965.1 - 7 - 8 1508 70

View Full Product Details