The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320653.1 | NP_001307582.1 | 5 - 6 | 945 | 119 |
NM_002532.5 | NP_002523.2 | 5 - 6 | 945 | 119 | |
XM_011523893.2 | XP_011522195.1 | 6 - 7 | 995 | 119 | |
XM_017024694.1 | XP_016880183.1 | 6 - 7 | 995 | 119 | |
GenBank mRNA | AK225247.1 | - | 5 - 6 | 865 | 119 |
AK298158.1 | - | 6 - 7 | 722 | 119 | |
AK299000.1 | - | 5 - 6 | 868 | 119 | |
AK308142.1 | - | 5 - 6 | 757 | 119 | |
BC000335.2 | - | 5 - 6 | 858 | 119 | |
Y08612.2 | - | 5 - 6 | 904 | 120 |