Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPATA6L
Entrez Gene ID
55064
Gene Name
spermatogenesis associated 6 like
Gene Aliases
C9orf68, bA6J24.2
Chromosome Location
Chr.9: 4540092 - 4666674 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC036349.1 - 1 - 2 96 81

View Full Product Details