The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015473.3 | NP_056288.2 | 17 - 18 | 2705 | 84 |
GenBank mRNA | AB037737.1 | - | 8 - 9 | 1153 | 84 |
AK092400.1 | - | 10 - 11 | 1447 | 84 | |
BC045764.1 | - | 11 - 12 | 1718 | 84 | |
CR749444.1 | - | 10 - 11 | 1403 | 84 |