Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAX4
Entrez Gene ID
5078
Gene Name
paired box 4
Gene Aliases
KPD, MODY9
Chromosome Location
Chr.7: 127610292 - 127618192 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB008913.1 - 8 - 9 915 65
BC107149.2 - 7 - 8 1518 65

View Full Product Details