The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004177.4 | NP_004168.1 | 10 - 11 | 1442 | 107 |
XM_005274200.3 | XP_005274257.1 | 9 - 10 | 1013 | 107 | |
XM_011545221.2 | XP_011543523.1 | 9 - 10 | 1024 | 107 | |
XM_017018188.1 | XP_016873677.1 | 10 - 11 | 886 | 107 | |
XM_017018195.1 | XP_016873684.1 | 9 - 10 | 775 | 107 | |
XM_017018196.1 | XP_016873685.1 | 8 - 9 | 916 | 107 | |
GenBank mRNA | AK125660.1 | - | 10 - 11 | 1442 | 107 |
AK290251.1 | - | 10 - 11 | 1011 | 107 | |
BC007405.2 | - | 10 - 11 | 1085 | 107 | |
BC007429.2 | - | 10 - 11 | 1085 | 107 | |
BX538158.1 | - | 11 - 12 | 1377 | 107 | |
U32315.1 | - | 10 - 11 | 933 | 107 |