The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004454.2 | NP_004445.1 | 9 - 10 | 1213 | 108 |
GenBank mRNA | AK301878.1 | - | 9 - 10 | 1128 | 108 |
BC007333.2 | - | 9 - 10 | 1176 | 108 | |
BT006713.1 | - | 8 - 9 | 966 | 108 | |
CR456767.1 | - | 8 - 9 | 966 | 108 | |
X76184.1 | - | 9 - 10 | 1161 | 108 |