Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LEKR1
Entrez Gene ID
389170
Gene Name
leucine, glutamate and lysine rich 1
Gene Aliases
-
Chromosome Location
Chr.3: 156825228 - 157047659 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004316.2 NP_001004316.2 5 - 6 670 162
XM_005247441.3 XP_005247498.1 5 - 6 2482 162
XM_006713631.3 XP_006713694.1 5 - 6 1194 162
XM_011512803.2 XP_011511105.1 5 - 6 1194 162
XM_011512804.2 XP_011511106.1 5 - 6 2715 162
XM_011512806.2 XP_011511108.1 5 - 6 1194 162
XM_011512807.2 XP_011511109.1 5 - 6 1194 162
XM_011512808.2 XP_011511110.1 5 - 6 1194 162
XM_011512809.1 XP_011511111.1 2 - 3 301 162
XM_011512811.2 XP_011511113.1 5 - 6 1194 162
GenBank mRNA AK131473.1 - 5 - 6 670 162

View Full Product Details