The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005262743.2 | XP_005262800.1 | 1 - 2 | 149 | 96 |
GenBank mRNA | DB040584.1 | - | 1 - 2 | 153 | 96 |
DB058857.1 | - | 1 - 2 | 114 | 96 |