Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OCEL1
Entrez Gene ID
79629
Gene Name
occludin/ELL domain containing 1
Gene Aliases
FWP009, S863-9
Chromosome Location
Chr.19: 17226204 - 17229219 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024578.2 NP_078854.1 1 - 2 112 145
XM_006722899.3 XP_006722962.1 1 - 2 111 145
GenBank mRNA AK026362.1 - 1 - 2 70 145
BC029361.1 - 1 - 2 112 145

View Full Product Details