The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016642.3 | NP_057726.4 | 4 - 5 | 734 | 85 |
XM_017022299.1 | XP_016877788.1 | 3 - 4 | 592 | 85 | |
XM_017022300.1 | XP_016877789.1 | 3 - 4 | 592 | 85 | |
XM_017022303.1 | XP_016877792.1 | 3 - 4 | 592 | 85 | |
XM_017022304.1 | XP_016877793.1 | 3 - 4 | 592 | 85 | |
XM_017022305.1 | XP_016877794.1 | 3 - 4 | 592 | 85 | |
GenBank mRNA | AF233523.1 | - | 4 - 5 | 734 | 85 |