The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004598.3 | NP_004589.1 | 9 - 10 | 1142 | 111 |
GenBank mRNA | AB209701.1 | - | 10 - 11 | 1126 | 111 |
AF231124.1 | - | 9 - 10 | 1012 | 111 | |
AK094489.1 | - | 8 - 9 | 1182 | 111 | |
BC030691.2 | - | 9 - 10 | 1123 | 111 |