Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FCHSD1
Entrez Gene ID
89848
Gene Name
FCH and double SH3 domains 1
Gene Aliases
NWK2
Chromosome Location
Chr.5: 141639302 - 141651453 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033449.2 NP_258260.1 12 - 13 1200 103
XM_005268524.4 XP_005268581.1 12 - 13 1227 103
XM_006714803.3 XP_006714866.1 11 - 12 1104 103
XM_011537698.2 XP_011536000.1 12 - 13 1233 103
XM_011537700.2 XP_011536002.1 12 - 13 1234 103
XM_011537701.2 XP_011536003.1 12 - 13 1239 103
XM_017010013.1 XP_016865502.1 12 - 13 1233 103
GenBank mRNA AK000007.1 - 12 - 13 1184 103
AY217346.1 - 12 - 13 1200 103
AY358478.1 - 3 - 4 818 103
BC047016.1 - 11 - 12 1127 103

View Full Product Details