The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033449.2 | NP_258260.1 | 17 - 18 | 1919 | 64 |
XM_005268524.4 | XP_005268581.1 | 17 - 18 | 1946 | 64 | |
XM_006714803.3 | XP_006714866.1 | 16 - 17 | 1823 | 64 | |
XM_017010013.1 | XP_016865502.1 | 17 - 18 | 1952 | 64 | |
GenBank mRNA | AK000007.1 | - | 17 - 18 | 1903 | 64 |
AY217346.1 | - | 17 - 18 | 1919 | 64 | |
AY358478.1 | - | 8 - 9 | 1537 | 64 |