Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDPK1
Entrez Gene ID
5170
Gene Name
3-phosphoinositide dependent protein kinase 1
Gene Aliases
PDK1, PDPK2, PDPK2P, PRO0461
Chromosome Location
Chr.16: 2537964 - 2603190 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002613.4 NP_002604.1 12 - 13 1551 56
NM_031268.5 NP_112558.2 9 - 10 1170 56
XM_011522521.2 XP_011520823.1 12 - 13 5239 56
XM_011522523.2 XP_011520825.1 9 - 10 4863 56
XM_017023271.1 XP_016878760.1 12 - 13 5244 56
XM_017023272.1 XP_016878761.1 12 - 13 1800 56
XM_017023273.1 XP_016878762.1 12 - 13 1509 56
XM_017023274.1 XP_016878763.1 13 - 14 1647 56
XM_017023275.1 XP_016878764.1 9 - 10 4869 56
XM_017023276.1 XP_016878765.1 9 - 10 1113 56
XM_017023277.1 XP_016878766.1 10 - 11 1211 56
GenBank mRNA AF017995.1 - 12 - 13 1482 56
AK131060.1 - 12 - 13 1485 56
AK222581.1 - 12 - 13 1494 56
BC006339.2 - 9 - 10 1131 56
BC012103.1 - 12 - 13 1511 56
BC033494.1 - 9 - 10 1083 56
CR536517.1 - 12 - 13 1402 56
Y15056.1 - 12 - 13 1481 56

View Full Product Details