Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRPA1
Entrez Gene ID
8989
Gene Name
transient receptor potential cation channel subfamily A member 1
Gene Aliases
ANKTM1, FEPS
Chromosome Location
Chr.8: 72021251 - 72094885 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007332.2 NP_015628.2 22 - 23 2857 92
XM_011517624.2 XP_011515926.1 23 - 24 7673 92
XM_011517625.2 XP_011515927.1 24 - 25 7832 92
XM_017013946.1 XP_016869435.1 23 - 24 4135 92
GenBank mRNA AK307942.1 - 22 - 23 2872 92
Y10601.1 - 22 - 23 2856 92

View Full Product Details