Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRPA1
Entrez Gene ID
8989
Gene Name
transient receptor potential cation channel subfamily A member 1
Gene Aliases
ANKTM1, FEPS
Chromosome Location
Chr.8: 72021251 - 72094885 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007332.2 NP_015628.2 26 - 27 3326 111
XM_011517624.2 XP_011515926.1 27 - 28 8142 111
XM_011517625.2 XP_011515927.1 28 - 29 8301 111
XM_017013946.1 XP_016869435.1 27 - 28 4604 111
GenBank mRNA Y10601.1 - 26 - 27 3325 111

View Full Product Details