Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SH3D19
Entrez Gene ID
152503
Gene Name
SH3 domain containing 19
Gene Aliases
EBP, EVE1, Kryn, SH3P19
Chromosome Location
Chr.4: 151120281 - 151228030 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009555.3 NP_001009555.3 19 - 20 3250 70
NM_001128923.1 NP_001122395.1 18 - 19 3169 70
NM_001128924.1 NP_001122396.1 11 - 12 2172 70
NM_001243349.1 NP_001230278.1 18 - 19 3090 70
XM_005262767.2 XP_005262824.1 20 - 21 3111 70
XM_011531646.1 XP_011529948.1 18 - 19 2897 70
XM_011531647.1 XP_011529949.1 17 - 18 2789 70
XM_011531648.1 XP_011529950.1 20 - 21 3224 70
XM_011531649.1 XP_011529951.1 16 - 17 2720 70
XM_017007778.1 XP_016863267.1 20 - 21 3215 70
XM_017007779.1 XP_016863268.1 16 - 17 2711 70
XM_017007780.1 XP_016863269.1 19 - 20 3146 70
XM_017007781.1 XP_016863270.1 19 - 20 3141 70
XM_017007782.1 XP_016863271.1 18 - 19 3072 70
XM_017007783.1 XP_016863272.1 19 - 20 3042 70
GenBank mRNA AK092528.1 - 11 - 12 1402 70
AK128493.1 - 11 - 12 2021 70
AK291025.1 - 11 - 12 1338 70
AK294476.1 - 18 - 19 3090 70
AL133047.1 - 9 - 10 1014 70
BC032468.1 - 1 - 2 103 70
BC045742.1 - 3 - 4 415 70
BC085613.1 - 10 - 11 1229 70
BC108890.1 - 12 - 13 2280 70
BC108891.1 - 12 - 13 2280 70
BC108892.1 - 12 - 13 2280 70
BC108893.1 - 11 - 12 2172 70
BX647422.1 - 19 - 20 3162 70

View Full Product Details