The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253837.1 | NP_001240766.1 | 8 - 9 | 1939 | 70 |
NM_016112.2 | NP_057196.2 | 8 - 9 | 1917 | 70 | |
XM_011540323.2 | XP_011538625.1 | 9 - 10 | 1480 | 70 | |
XM_011540325.2 | XP_011538627.1 | 10 - 11 | 1590 | 70 | |
XM_017016875.1 | XP_016872364.1 | 9 - 10 | 1428 | 70 | |
XM_017016876.1 | XP_016872365.1 | 7 - 8 | 1186 | 70 | |
GenBank mRNA | AF053316.1 | - | 8 - 9 | 1409 | 70 |
AF073481.1 | - | 8 - 9 | 1917 | 70 | |
AF092170.1 | - | 6 - 7 | 1196 | 70 | |
AF094827.1 | - | 7 - 8 | 1249 | 70 | |
BC025665.1 | - | 8 - 9 | 1614 | 70 | |
DQ084244.1 | - | 8 - 9 | 1393 | 70 |