Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATG13
Entrez Gene ID
9776
Gene Name
autophagy related 13
Gene Aliases
KIAA0652, PARATARG8
Chromosome Location
Chr.11: 46617276 - 46676019 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142673.2 NP_001136145.1 12 - 13 1578 86
NM_001205119.1 NP_001192048.1 12 - 13 1621 86
NM_001205120.1 NP_001192049.1 11 - 12 1522 86
XM_005253262.2 XP_005253319.1 13 - 14 1417 86
XM_005253263.2 XP_005253320.1 13 - 14 1708 86
XM_005253265.2 XP_005253322.1 14 - 15 1914 86
XM_005253266.2 XP_005253323.1 13 - 14 1643 86
XM_005253268.2 XP_005253325.1 11 - 12 1020 86
XM_006718394.2 XP_006718457.1 14 - 15 1764 86
XM_006718395.2 XP_006718458.1 14 - 15 1699 86
XM_006718396.2 XP_006718459.1 13 - 14 1798 86
XM_011520493.1 XP_011518795.1 15 - 16 1901 86
XM_011520496.2 XP_011518798.1 12 - 13 1676 86
XM_011520499.2 XP_011518801.1 12 - 13 1290 86
XM_011520500.1 XP_011518802.1 14 - 15 1737 86
XM_011520501.1 XP_011518803.1 13 - 14 1665 86
XM_011520502.1 XP_011518804.1 13 - 14 1638 86
XM_017018599.1 XP_016874088.1 14 - 15 1849 86
XM_017018600.1 XP_016874089.1 13 - 14 1390 86
XM_017018601.1 XP_016874090.1 12 - 13 1565 86
XM_017018602.1 XP_016874091.1 13 - 14 1815 86
XM_017018603.1 XP_016874092.1 14 - 15 1932 86
XM_017018604.1 XP_016874093.1 12 - 13 1609 86
XM_017018605.1 XP_016874094.1 13 - 14 1600 86
XM_017018606.1 XP_016874095.1 12 - 13 1544 86
XM_017018607.1 XP_016874096.1 10 - 11 921 86
XM_017018608.1 XP_016874097.1 12 - 13 1291 86
XM_017018609.1 XP_016874098.1 12 - 13 1582 86
XM_017018610.1 XP_016874099.1 11 - 12 1466 86
XM_017018611.1 XP_016874100.1 12 - 13 1517 86
XM_017018618.1 XP_016874107.1 14 - 15 1903 86
XM_017018619.1 XP_016874108.1 12 - 13 1688 86
XM_017018620.1 XP_016874109.1 12 - 13 1748 86
XM_017018621.1 XP_016874110.1 11 - 12 1632 86
GenBank mRNA AB014552.1 - 11 - 12 1202 86
AK223603.1 - 12 - 13 1297 86
AK289641.1 - 12 - 13 1125 86
AK296526.1 - 4 - 5 409 86
AK315925.1 - 4 - 5 336 86

View Full Product Details