Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATG13
Entrez Gene ID
9776
Gene Name
autophagy related 13
Gene Aliases
KIAA0652, PARATARG8
Chromosome Location
Chr.11: 46617276 - 46676019 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142673.2 NP_001136145.1 16 - 17 2034 77
NM_001205119.1 NP_001192048.1 16 - 17 2077 77
NM_001205120.1 NP_001192049.1 15 - 16 1978 77
NM_001205121.1 NP_001192050.1 15 - 16 1918 77
NM_001205122.1 NP_001192051.1 13 - 14 1785 77
NM_014741.4 NP_055556.2 15 - 16 1923 77
XM_005253262.2 XP_005253319.1 17 - 18 1873 77
XM_005253263.2 XP_005253320.1 17 - 18 2164 77
XM_005253265.2 XP_005253322.1 18 - 19 2370 77
XM_005253266.2 XP_005253323.1 17 - 18 2099 77
XM_005253268.2 XP_005253325.1 15 - 16 1476 77
XM_006718394.2 XP_006718457.1 18 - 19 2220 77
XM_006718395.2 XP_006718458.1 18 - 19 2155 77
XM_006718396.2 XP_006718459.1 17 - 18 2254 77
XM_011520493.1 XP_011518795.1 19 - 20 2357 77
XM_011520496.2 XP_011518798.1 16 - 17 2132 77
XM_011520499.2 XP_011518801.1 16 - 17 1746 77
XM_011520500.1 XP_011518802.1 18 - 19 2193 77
XM_011520501.1 XP_011518803.1 17 - 18 2121 77
XM_011520502.1 XP_011518804.1 17 - 18 2094 77
XM_011520503.1 XP_011518805.1 16 - 17 2010 77
XM_017018599.1 XP_016874088.1 18 - 19 2305 77
XM_017018600.1 XP_016874089.1 17 - 18 1846 77
XM_017018601.1 XP_016874090.1 16 - 17 2021 77
XM_017018602.1 XP_016874091.1 17 - 18 2271 77
XM_017018603.1 XP_016874092.1 18 - 19 2388 77
XM_017018604.1 XP_016874093.1 16 - 17 2065 77
XM_017018605.1 XP_016874094.1 17 - 18 2056 77
XM_017018606.1 XP_016874095.1 16 - 17 2000 77
XM_017018607.1 XP_016874096.1 14 - 15 1377 77
XM_017018608.1 XP_016874097.1 16 - 17 1747 77
XM_017018609.1 XP_016874098.1 16 - 17 2038 77
XM_017018610.1 XP_016874099.1 15 - 16 1922 77
XM_017018611.1 XP_016874100.1 16 - 17 1973 77
XM_017018612.1 XP_016874101.1 16 - 17 2160 77
XM_017018613.1 XP_016874102.1 15 - 16 1954 77
XM_017018614.1 XP_016874103.1 16 - 17 1945 77
XM_017018615.1 XP_016874104.1 15 - 16 2044 77
XM_017018616.1 XP_016874105.1 14 - 15 1838 77
XM_017018617.1 XP_016874106.1 16 - 17 2043 77
XM_017018618.1 XP_016874107.1 18 - 19 2359 77
XM_017018619.1 XP_016874108.1 16 - 17 2144 77
XM_017018620.1 XP_016874109.1 16 - 17 2204 77
XM_017018621.1 XP_016874110.1 15 - 16 2088 77
XM_017018622.1 XP_016874111.1 15 - 16 2045 77
GenBank mRNA AB014552.1 - 15 - 16 1658 77
AK023867.1 - 15 - 16 1656 77
AK223603.1 - 16 - 17 1753 77
AK289641.1 - 16 - 17 1581 77
AK293109.1 - 15 - 16 1622 77
AK294110.1 - 13 - 14 1498 77
AK296526.1 - 8 - 9 865 77
AK315925.1 - 8 - 9 792 77
BC001331.1 - 15 - 16 1352 77
BC002378.2 - 15 - 16 1622 77

View Full Product Details