The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278192.1 | NP_001265121.1 | 20 - 21 | 2481 | 61 |
NM_001278193.1 | NP_001265122.1 | 19 - 20 | 2415 | 61 | |
NM_001278194.1 | NP_001265123.1 | 21 - 22 | 2648 | 61 | |
NM_198334.2 | NP_938148.1 | 22 - 23 | 2757 | 61 | |
NM_198335.3 | NP_938149.2 | 23 - 24 | 2823 | 61 | |
XM_017017412.1 | XP_016872901.1 | 20 - 21 | 2491 | 61 | |
XM_017017413.1 | XP_016872902.1 | 22 - 23 | 2873 | 61 | |
XM_017017414.1 | XP_016872903.1 | 22 - 23 | 2704 | 61 | |
GenBank mRNA | AF144074.1 | - | 23 - 24 | 2712 | 61 |
AJ000332.1 | - | 22 - 23 | 2630 | 61 | |
AK026548.1 | - | 11 - 12 | 1161 | 61 | |
AK026567.1 | - | 3 - 4 | 299 | 61 | |
AK026997.1 | - | 22 - 23 | 2648 | 61 | |
AK295806.1 | - | 19 - 20 | 2303 | 61 | |
AK299819.1 | - | 21 - 22 | 2531 | 61 | |
AK302752.1 | - | 20 - 21 | 2387 | 61 | |
AK314370.1 | - | 22 - 23 | 2646 | 61 | |
BC005405.1 | - | 5 - 6 | 737 | 61 | |
BC017433.2 | - | 12 - 13 | 1262 | 61 | |
BC017435.2 | - | 12 - 13 | 1262 | 61 | |
BC034439.1 | - | 13 - 14 | 1633 | 61 | |
BC065266.1 | - | 21 - 22 | 2524 | 61 | |
D42041.1 | - | 22 - 23 | 2628 | 61 | |
EU794613.1 | - | 22 - 23 | 2663 | 61 |