The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278192.1 | NP_001265121.1 | 6 - 7 | 665 | 88 |
NM_001278193.1 | NP_001265122.1 | 5 - 6 | 599 | 88 | |
NM_001278194.1 | NP_001265123.1 | 7 - 8 | 832 | 88 | |
NM_198334.2 | NP_938148.1 | 8 - 9 | 941 | 88 | |
NM_198335.3 | NP_938149.2 | 9 - 10 | 1007 | 88 | |
XM_017017412.1 | XP_016872901.1 | 6 - 7 | 675 | 88 | |
XM_017017413.1 | XP_016872902.1 | 8 - 9 | 1057 | 88 | |
XM_017017414.1 | XP_016872903.1 | 8 - 9 | 888 | 88 | |
GenBank mRNA | AF144074.1 | - | 9 - 10 | 896 | 88 |
AJ000332.1 | - | 8 - 9 | 814 | 88 | |
AK026997.1 | - | 8 - 9 | 831 | 88 | |
AK299819.1 | - | 7 - 8 | 715 | 88 | |
AK314370.1 | - | 8 - 9 | 830 | 88 | |
BC065266.1 | - | 7 - 8 | 708 | 88 | |
D42041.1 | - | 8 - 9 | 812 | 88 | |
EU794613.1 | - | 8 - 9 | 847 | 88 |