Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VIP
Entrez Gene ID
7432
Gene Name
vasoactive intestinal peptide
Gene Aliases
PHM27
Chromosome Location
Chr.6: 152747894 - 152759765 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003381.3 NP_003372.1 5 - 6 635 94
NM_194435.2 NP_919416.1 5 - 6 632 94
GenBank mRNA AK313950.1 - 5 - 6 635 94
BC009794.1 - 5 - 6 637 94
M36634.1 - 5 - 6 528 94
M54930.1 - 3 - 4 315 94

View Full Product Details