Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM173A
Entrez Gene ID
65990
Gene Name
family with sequence similarity 173 member A
Gene Aliases
C16orf24
Chromosome Location
Chr.16: 720581 - 722590 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271285.1 NP_001258214.1 1 - 2 298 95
NM_023933.2 NP_076422.1 1 - 2 298 95
GenBank mRNA BC001181.1 - 1 - 2 282 95
BC002624.2 - 1 - 2 250 95
BI597021.1 - 1 - 2 303 95
BI838160.1 - 1 - 2 281 95

View Full Product Details