The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005359.5 | NP_005350.1 | 3 - 4 | 967 | 77 |
GenBank mRNA | AK290770.1 | - | 3 - 4 | 629 | 77 |
BC002379.2 | - | 4 - 5 | 662 | 77 | |
KU178168.1 | - | 2 - 3 | 429 | 77 | |
KU178169.1 | - | 2 - 3 | 429 | 77 | |
U44378.1 | - | 2 - 3 | 557 | 77 |