Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ANAPC15
- Entrez Gene ID
-
25906
- Gene Name
- anaphase promoting complex subunit 15
- Gene Aliases
-
APC15, C11orf51, HSPC020
- Chromosome Location
-
Chr.11: 72106254 - 72112793 on Build GRCh38
View Full Product Details