The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282538.1 | NP_001269467.1 | 7 - 8 | 943 | 64 |
NM_001282539.1 | NP_001269468.1 | 8 - 9 | 1040 | 64 | |
NM_002074.4 | NP_002065.1 | 9 - 10 | 1089 | 64 | |
XM_017001059.1 | XP_016856548.1 | 8 - 9 | 993 | 64 | |
XM_017001060.1 | XP_016856549.1 | 9 - 10 | 863 | 64 | |
XM_017001061.1 | XP_016856550.1 | 8 - 9 | 1036 | 64 | |
GenBank mRNA | AF070597.1 | - | 1 - 2 | 58 | 64 |
AF070603.1 | - | 3 - 4 | 297 | 64 | |
AK122949.1 | - | 7 - 8 | 727 | 64 | |
AK312614.1 | - | 10 - 11 | 851 | 64 | |
BC004186.2 | - | 9 - 10 | 972 | 64 | |
BC005888.2 | - | 7 - 8 | 740 | 64 | |
BC008991.1 | - | 9 - 10 | 1028 | 64 | |
BC114618.1 | - | 8 - 9 | 811 | 64 | |
BT007305.1 | - | 7 - 8 | 696 | 64 | |
CR456784.1 | - | 7 - 8 | 696 | 64 | |
M36430.1 | - | 3 - 4 | 417 | 64 |