The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168335.1 | NP_001161807.1 | 10 - 11 | 1312 | 111 |
NM_002396.4 | NP_002387.1 | 10 - 11 | 1312 | 111 | |
GenBank mRNA | AK313391.1 | - | 10 - 11 | 1208 | 111 |
BC000147.1 | - | 10 - 11 | 1254 | 111 | |
M55905.1 | - | 10 - 11 | 1142 | 111 |