The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153367.3 | NP_699198.2 | 2 - 3 | 636 | 106 |
XM_011539452.2 | XP_011537754.1 | 2 - 3 | 587 | 106 | |
GenBank mRNA | AK075279.1 | - | 2 - 3 | 488 | 106 |
AK293361.1 | - | 2 - 3 | 275 | 106 | |
AK303734.1 | - | 2 - 3 | 267 | 106 | |
BC028617.1 | - | 2 - 3 | 638 | 106 | |
BC053564.1 | - | 2 - 3 | 535 | 106 |