The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006835.2 | NP_006826.1 | 5 - 6 | 1007 | 91 |
GenBank mRNA | AB209766.1 | - | 5 - 6 | 1009 | 91 |
AF135162.1 | - | 5 - 6 | 662 | 91 | |
AK222931.1 | - | 4 - 5 | 459 | 91 | |
AK301111.1 | - | 4 - 5 | 868 | 91 | |
AK312632.1 | - | 7 - 8 | 877 | 91 | |
BC000420.2 | - | 5 - 6 | 1006 | 91 | |
BC004975.1 | - | 5 - 6 | 1006 | 91 | |
BT020019.1 | - | 4 - 5 | 463 | 91 | |
CR541783.1 | - | 4 - 5 | 463 | 91 | |
D50310.1 | - | 4 - 5 | 463 | 91 |